Learn More
Medchemexpress LLC 4'-Demethylepipodophyllotoxin | 6559-91-7 | 99.8% | 400.38 | 5 G

Supplier: Medchemexpress LLC HY174355G
4'-Demethylepipodophyllotoxin is a solid chemical supplied by MedChemExpress for laboratory use and the manufacture of substances. This compound has a molecular weight of 400.38 and a purity of 99.76% by HPLC. It is stable under recommended storage conditions, but appropriate handling precautions should be observed due to its classification as possibly carcinogenic and reproductively toxic.
- Appears as a white to off-white solid
- Melting/freezing point between 246-248 °C
- Boiling point/range is 626.5±55.0 °C at 760 mmHg
- Store powder at -20°C for 3 years, or 4°C for 2 years
- Store in solvent at -80°C for 2 years, or -20°C for 1 year
- Wear protective gloves, clothing, eye, and face protection
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.