Learn More
Medchemexpress LLC 4′-Demethylnobiletin | 34810-62-3 | 99.8% | 388.37 g/mol | C20H20O8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1420665MG
Encompass_Preferred
4′-Demethylnobiletin is a bioactive flavonoid metabolite used as a research reagent to modulate intracellular signaling in neuroscience and biochemical studies. It activates PKA/ERK/CREB pathways, enhances CRE-mediated transcription in hippocampal neurons, and has been reported to reverse memory impairment associated with NMDA receptor antagonism. The compound is supplied at high purity for laboratory assays.
  • Activates PKA/ERK/CREB signaling pathways
  • Enhances CRE-mediated transcription in hippocampal neurons
  • Reverses NMDA receptor antagonist-related memory impairment in models
  • High purity suitable for biochemical assays (99.8%)
  • Available in small-quantity formats for laboratory research
  • Molecular formula C20H20O8; molecular weight 388.37 g/mol
Catalog No. 50-002-21600