Learn More
Selleck Chemical LLC 4-Demethylpodophyllotoxin S3908-25mg
SDP
Supplier:  Selleck Chemical LLC S390825MG
Encompass_Preferred
4 -Demethylpodophyllotoxin is isolated from the herb of Dysosma pleiantha (Hance) Woodson with remarkable cytotoxic potential in diverse cancer cell lines
Catalog No. 50-311-0567