Learn More
Medchemexpress LLC 4-desmethoxy omeprazole | 110374-16-8 | MFCD08272115 | 99.0% | 315.39 | C16H17N3O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1351115MG
Encompass_Preferred
4-Desmethoxy Omeprazole is the active metabolite and analytical standard of omeprazole, intended for research and analytical applications including impurity profiling, metabolite studies, and method development. The material is provided as a characterized solid reference with reported high purity and defined molecular properties.
  • High purity analytical standard suitable for chromatography and mass spectrometry.
  • Intended for impurity identification, metabolite studies, and method validation.
  • Available in small milligram-scale quantities appropriate for laboratory use.
  • Supplied with CAS, molecular formula, and molecular weight information for traceability.
Catalog No. 50-002-23470