Learn More
Medchemexpress LLC 4-(dimethylamino)cinnamaldehyde | 6203-18-5 | MFCD00007002 | 99.8% | 175.23 g/mol | C11H13NO | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW016729
Encompass_Preferred
4-(Dimethylamino)cinnamaldehyde is an aromatic α,β-unsaturated aldehyde reagent used to detect indole-containing compounds produced by tryptophan decarboxylase and tryptophanase and as a synthetic intermediate in organic chemistry. It is supplied as a high-purity solid suitable for biochemical assays and analytical applications.
Catalog No. 50-003-02761