Learn More
Medchemexpress LLC 4-(dimethylamino)phenacyl bromide-d6 | 37904-72-6 | 98.8% | 248.15 g/mol | C10H6D6BrNO | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW128417S15MG
Encompass_Preferred
4-(Dimethylamino)phenacyl bromide-d6 is a deuterium-labeled reagent provided for research use as a tracer and internal standard in quantitative analytical methods. It delivers defined isotopic labeling and high chemical purity to support accurate quantification in small-scale analytical workflows.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High purity and defined isotopic labeling.
  • Available in small mg-scale quantities for analytical use.
  • Intended for research use only; not for clinical or diagnostic use.
Catalog No. 50-002-82811