Learn More
Medchemexpress LLC 4-dimethylaminopyridine N-oxide | 1005-31-8 | MFCD00143310 | 98.0% | 138.17 g/mol | C7H10N2O | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW145090250MG
Encompass_Preferred
4-Dimethylaminopyridine N-oxide is an oxidizing reagent used in organic synthesis, particularly for the oxidation of alkyl halides. It is supplied as a solid, research-grade chemical (CAS 1005-31-8) with a molecular weight of 138.17 g/mol and is intended for laboratory research and method development.
  • Efficient oxidant for alkyl halides
  • Solid form suitable for bench-scale handling
  • Available in small pack sizes for research use
  • Commonly used in synthetic method development
  • Provided for research use only
Catalog No. 50-002-83743