Learn More
Medchemexpress LLC 4-dodecyl-N-(1,3,4-thiadiazol-2-yl)benzenesulfonamide | 1191951-57-1 | 98.1% | 5 MG

Supplier: Medchemexpress LLC HY120635MG
PHT-427 is a small-molecule research inhibitor that targets the pleckstrin homology (PH) domains of Akt and PDPK1 and is used to probe Akt/PDPK1 signaling in cellular and in vivo studies.
- Inhibits Akt PH domain (Ki = 2.7 μM).
- Inhibits PDPK1 PH domain (Ki = 5.2 μM).
- Molecular formula C20H31N3O2S2; molecular weight 409.61.
- High purity (98.1% by HPLC).
- CAS number 1191951-57-1.
- Typical supply: 5 mg solid.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.