Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-epi-oxytetracycline | 14206-58-7 | ≥95.0% | 460.43 | C22H24N2O9 | 10MG
SDP

Supplier:  Medchemexpress LLC HY12594710MG

Encompass_Preferred

4-Epioxytetracycline is a degradation product and metabolite of the antibiotic oxytetracycline supplied as an analytical standard for environmental and pharmaceutical analysis. It is used for qualitative and quantitative assays, method development, and trace-level monitoring by chromatographic and mass-spectrometric techniques.

  • Analytical standard for 4-epioxytetracycline, a metabolite of oxytetracycline.
  • CAS number 14206-58-7.
  • Molecular formula C22H24N2O9; molecular weight 460.43 g/mol.
  • Purity ≥95.0% (batch-specific; see certificate of analysis).
  • Supplied as a solid/powder; common pack sizes include 50 mg, 100 mg, and 250 mg (other sizes may be available).
  • Intended applications: HPLC, GC, MS, quantitative and qualitative method development.
  • Refer to the certificate of analysis or product documentation for storage and batch-specific specifications.

Catalog No. 50-002-35344


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.