Learn More
Medchemexpress LLC 4-ethyloctanoic acid | 16493-80-4 | MFCD00506494 | 99.1% | 172.27 g/mol | C10H20O2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW015307100G
Encompass_Preferred
4-ethyloctanoic acid (CAS 16493-80-4) is a medium-chain branched carboxylic acid naturally occurring in plant sources and used as a flavoring compound and food additive. Supplied as a high-purity research chemical, it is suitable for analytical studies, formulation development, and sensory evaluation work.
  • High purity suitable for analytical use.
  • Distinct fatty odor and flavor characteristics used in sensory profiling.
  • Soluble in DMSO and compatible with common in vivo vehicles.
  • Stable when stored under recommended temperature conditions.
  • Available in multiple pack sizes for laboratory needs.
Catalog No. 50-003-03672