Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-ethyloctanoic acid | 16493-80-4 | MFCD00506494 | 99.1% | 172.27 g·mol⁻¹ | C10H20O2 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW015307

Encompass_Preferred

4-Ethyloctanoic acid is a naturally occurring carboxylic acid (octanoic acid, 4-ethyl-) used as a flavoring compound and food additive. It is supplied as a research-grade chemical with documented purity, molecular data, and solubility information suitable for formulation, analytical reference, and flavor chemistry studies.

  • High purity: 99.05%.
  • Molecular formula: C10H20O2.
  • Molecular weight: 172.27 g·mol⁻¹.
  • Soluble in DMSO at 100 mg/mL; sonication may be required.
  • In vivo formulation protocols report solubility ≥ 2.5 mg/mL.
  • Available in multiple pack sizes including 25 g.

Catalog No. 50-003-03069


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.