Learn More
Medchemexpress LLC 4-ethyloctanoic acid | 16493-80-4 | MFCD00506494 | 99.1% | 172.27 g/mol | C10H20O2 | 50 G

Supplier: Medchemexpress LLC HYW01530750G
4-Ethyloctanoic acid is a naturally occurring fatty acid first isolated from Saussurea lappa Clarke. It is supplied for laboratory and research use and is commonly used as a flavoring compound and food additive.
- Natural compound originally isolated from Saussurea lappa Clarke.
- Used as a flavoring compound and food additive.
- High purity (99.05%) suitable for analytical and research applications.
- Chemical formula C10H20O2; molecular weight 172.27 g/mol.
- Available in multiple pack sizes, including 50 g, for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.