Learn More
Medchemexpress LLC 4-Ethylphenylacetylene | 40307-11-7 | 98.11% | 130.19 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW012717100G
Encompass_Preferred
1-Ethyl-4-ethynylbenzene is a biochemical reagent suitable for life science research as a biological material or organic compound. It functions as a click chemistry reagent, featuring an Alkyne group that can engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Azide groups.
  • Biochemical reagent for life science research.
  • Click chemistry reagent.
  • Contains an alkyne group for CuAAc reactions.
Catalog No. 50-004-54824