Learn More
Medchemexpress LLC 4-Ethynylanisole | 768-60-5 | 132.16 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW004104
Encompass_Preferred
4-Ethynylanisole is a biochemical reagent used as a biological material or organic compound for life science-related research. It is a click chemistry reagent containing an Alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. In vitro, it has been used in the synthesis of photoluminescent 1,2-dihydrophosphinines via a [4+2] cycloaddition.
  • Biochemical reagent for life science research
  • Click chemistry reagent with Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Used in synthesis of photoluminescent 1,2-dihydrophosphinines via [4+2] cycloaddition
Catalog No. 50-004-48277