Learn More
Medchemexpress LLC 4-Ethynylanisole | 768-60-5 | MFCD00168815 | 132.16 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00410410G
Encompass_Preferred
4-Ethynylanisole is a biochemical reagent used in life science research. It functions as a click chemistry reagent containing an alkyne group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. It has been utilized in the synthesis of photoluminescent 1,2-dihydrophosphinines via a [4+2] cycloaddition.
  • Biochemical reagent for life science research
  • Click chemistry reagent with alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Used in synthesis of photoluminescent 1,2-dihydrophosphinines
Catalog No. 50-004-45877