Learn More
Medchemexpress LLC 4-Ethynylbenzoic acid | 10602-00-3 | 146.15 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00753310G
Encompass_Preferred
4-Ethynylbenzoic acid is a biochemical reagent used in life science research. It serves as a click chemistry reagent, containing an Alkyne group that enables it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Azide groups.
  • Biochemical reagent for life science-related research
  • Functions as a click chemistry reagent
  • Contains an Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups
Catalog No. 50-004-45805