Learn More
Medchemexpress LLC 4-Fluoro-2-methoxy-5-nitroaniline | 1075705-01-9 | 99.7% | C7H7FN2O3 | 100 G

Supplier: Medchemexpress LLC HY78863100G
4-Fluoro-2-Methoxy-5-Nitroaniline is a drug intermediate used for the synthesis of various active compounds. It is sold for research purposes only and not for patient use. The product appears as a solid with a color ranging from yellow to orange.
- Molecular weight: 186.14
- Appearance: Solid
- Color: Yellow to orange
- Storage: 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
- Shipping: Room temperature in continental US; may vary elsewhere.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.