Learn More
Medchemexpress LLC 4-Fluoro-2-Methoxy-5-Nitroaniline | 1075705-01-9 | 99.74% | C7H7FN2O3 | 10 G

Supplier: Medchemexpress LLC HY7886310G
4-Fluoro-2-Methoxy-5-Nitroaniline is a drug intermediate for the synthesis of various active compounds.
- Purity: 99.74%
- Molecular weight: 186.14
- Formula: C7H7FN2O3
- CAS no.: 1075705-01-9
- Appearance: Solid
- Color: Yellow to orange
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.