Learn More
Medchemexpress LLC 4-Fluoro-2-Methoxy-5-Nitroaniline | 1075705-01-9 | MFCD23098941 | C7H7FN2O3 | 500 G
SDP
Supplier:  Medchemexpress LLC HY78863500G
Encompass_Preferred
4-Fluoro-2-Methoxy-5-Nitroaniline is a chemical compound primarily used as a laboratory chemical and for the manufacture of other substances. This product is intended for research use only, and its chemical, physical, and toxicological properties have not been completely investigated.
  • Harmful if swallowed.
  • Causes skin irritation.
  • Causes serious eye irritation.
  • May cause respiratory irritation.
  • Store in a cool, well-ventilated area.
  • Protect from light.
Catalog No. 50-004-32068