Learn More
Medchemexpress LLC 4-Fluoro-5-hydroxy-2-methylindole | 288385-88-6 | 98.88% | C9H8FNO | 5 G
SDP
Supplier:  Medchemexpress LLC HY652685G
Encompass_Preferred
4-Fluoro-5-hydroxy-2-methylindole is a drug intermediate for the synthesis of various active compounds. It is supplied as a white to off-white solid with a purity of 98.88%.
  • Drug intermediate for synthesis of active compounds
  • Available in multiple package sizes
  • Solid appearance
  • White to off-white color
  • For research use only
Catalog No. 50-004-34450