Learn More
Medchemexpress LLC 4-Fluoro-5-hydroxy-2-methylindole | 288385-88-6 | 98.9% | C9H8FNO | 1 G

Supplier: Medchemexpress LLC HY652681G
4-Fluoro-5-hydroxy-2-methylindole is a drug intermediate used for the synthesis of various active compounds. This product is for research use only and not for sale to patients.
- Purity: 98.9%
- Molecular weight: 165.17
- Chemical formula: C9H8FNO
- Appearance: Solid
- Color: White to off-white
- Storage: 4°C, stored under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month (both under nitrogen).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.