Learn More
Medchemexpress LLC 4-Fluoro-5-hydroxy-2-methylindole | 288385-88-6 | 98.9% | C9H8FNO | 10 G

Supplier: Medchemexpress LLC HY6526810G
4-Fluoro-5-hydroxy-2-methylindole is a drug intermediate used for the synthesis of various active compounds. This product appears as a solid, white to off-white in color, and is intended for research use only.
- Used for the synthesis of various active compounds
- Appears as a solid
- Color is white to off-white
- For research use only
- Storage at 4°C, stored under nitrogen
- When in solvent, store at -80°C for 6 months or -20°C for 1 month, stored under nitrogen
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.