Learn More
Sigma Aldrich Fine Chemicals Biosciences 4 fluoro 7 sulfamoylbenzof10MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences F363910MG
Encompass
4-fluoro-7-sulfamoylbenzofurazan is a fluorescent reagent which directly links to the sulfur atom of thiols without a flexible alkyl chain. It is used to detect low molecular thiols using high performance liquid chromatography.
Catalog No. 50-290-2158