Learn More
Medchemexpress LLC 4-fluoro-N-[1-[1-(oxane-4-carbonyl)-2,3-dihydroindol-5-yl]ethyl]benzamide | 2166616-74-4 | ≥98% | 396.45 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY111540100MG
Encompass_Preferred
LY-3381916 (IDO1-IN-5) is a potent, selective, and brain-penetrant inhibitor of IDO1 activity. It functions by binding to apo-IDO1, which lacks heme, rather than to mature heme-bound IDO1.
  • Potent IDO1 inhibitor
  • Selective IDO1 inhibitor
  • Brain-penetrant IDO1 inhibitor
  • Binds to apo-IDO1 lacking heme
  • Does not show obvious agonism of aryl hydrocarbon receptor (AHR) at concentrations up to 100 μM.
Catalog No. 50-003-46000