Learn More
Medchemexpress LLC 4-fluoroacetophenone | 403-42-9 | MFCD00000354 | 99.9% | 138.14 | C8H7FO | 100g
SDP
Supplier:  Medchemexpress LLC HYY0779100G
Encompass_Preferred
4 -Fluoroacetophenone (1-(4-Fluorophenyl)ethan-1-one) is a biochemical reagent that can be used as a biological material or organic compound for life science related research 4 -Fluoroacetophenone is also an important intermediate that can be used in the synthesis of other active compounds such as aromatic diketones[1][2]
Catalog No. 50-002-69833