Learn More
Medchemexpress LLC 4-Formylcolchicine | 2730-82-7 | 99.8% | 427.45 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14734810MM/1ML
Encompass_Preferred
4-Formylcolchicine is a potent anticancer agent. It exhibits cytotoxicity activity with IC50 values of 1.007 μM for A549 cells, 0.128 μM for HT-29 cells, and 0.054 μM for HCT116 cells.
  • Potent anticancer agent
  • Exhibits cytotoxicity in A549, HT-29, and HCT116 cells
  • Soluble in DMSO (100 mg/mL)
  • Solid storage at 4°C, sealed, away from moisture and light
  • In-solvent storage at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light
Catalog No. 50-003-62061