Learn More
Cayman Chemical 4-HexylresorcInol 100g
SDP
Supplier:  Cayman Chemical 34824100 G
Encompass_Preferred
An alkylresorcinol; active against S. aureus, S. pyogenes, M. catarrhalis, H. influenzae, and F. necrophorum in an in vitro model of oral transmucosal delivery using 4-hexylresorcinol-containing lozenges; increases the activities of GPX, GR, and catalase in isolated human peripheral blood lymphocytes at 25, 50, or 100 µM; reduces tumor growth and increases survival in a PDX mouse model of oral mucosal melanoma at 2.5 mg/kg in combination with cisplatin
Catalog No. 50-291-1064