Learn More
Selleck Chemical LLC 4-HNE S9793-100MG
SDP
Supplier:  Selleck Chemical LLC S9793100MG
Encompass_Preferred
4-HNE (4-Hydroxynonenal) is one of the major end products of lipid peroxidation and has been widely accepted as an inducer of oxidative stress
Catalog No. 50-000-24586