Learn More
Medchemexpress LLC HY-W091541 10mg Medchemexpress, 4-Hydroxy-1H-indole-3-carbaldehyde CAS:81779-27-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW09154110MG
Encompass_Preferred
Medchemexpress, HY-W091541 10mg 4-Hydroxy-1H-indole-3-carbaldehyde CAS:81779-27-3 4-Hydroxy-1H-indole-3-carbaldehyde is a plant metabolite found in Capparis spinosa L.. 4-Hydroxy-1H-indole-3-carbaldehyde can be used in the synthesis of fluorescent probe. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3512