Learn More
Medchemexpress LLC 4-hydroxy-1H-indole-3-carbaldehyde | 81779-27-3 | MFCD05864721 | 98.1% | 161.16 g/mol | C9H7NO2 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW091541
4-Hydroxy-1H-indole-3-carbaldehyde is a plant-derived small molecule used as a research reagent and synthetic intermediate. It is applied in the preparation of fluorescent probes and in studies of inflammation-related biological pathways.
- Purity 98.1%.
- Molecular formula C9H7NO2.
- Molecular weight 161.16 g/mol.
- CAS number 81779-27-3.
- Available packaging: 10 mg, 25 mg, 50 mg, 100 mg, 250 mg, 500 mg, 1 g, 5 g.
- Suitable for chemical synthesis and biochemical research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.