Learn More
Medchemexpress LLC 4-hydroxy-3,3-dimethyl-2H-benz[g]indole-2,5(3H)-dione | 39674-97-0 | MFCD00616487 | 98.4% | 241.24 g/mol | C14H11NO3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10062550MG
Encompass_Preferred
BVT948 is a small-molecule research compound that functions as a protein tyrosine phosphatase (PTP) inhibitor. It is used in biochemical and cell-based studies to probe PTP-mediated signaling and has reported activity against several cytochrome P450 isoforms and the lysine methyltransferase SETD8.
  • Noncompetitive, irreversible PTP inhibition with a cell-permeable profile.
  • Also inhibits multiple cytochrome P450 isoforms and SETD8.
  • High purity (98.4%) suitable for biochemical and cellular assays.
  • Solid, pink to red appearance; available as solid material and as DMSO solution.
  • Recommended sealed storage to preserve stability in solid or solution form.
Catalog No. 50-002-14269