Learn More
Medchemexpress LLC 4-hydroxy-3,3-dimethyl-2H-benzo[g]indole-2,5(3H)-dione | 39674-97-0 | MFCD00616487 | 98.4% | 241.24 g·mol⁻¹ | C14H11NO3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10062510MG
Encompass_Preferred
BVT948 is a small-molecule research compound that inhibits protein tyrosine phosphatases (PTPs) and shows activity against multiple cytochrome P450 isoforms and the lysine methyltransferase SETD8 (KMT5A). It is supplied as a solid for biochemical and cell-based assays and is typically provided in small research pack sizes for laboratory use.
  • Inhibits protein tyrosine phosphatases (PTPs).
  • Shows activity against several cytochrome P450 isoforms.
  • Inhibits lysine methyltransferase SETD8 (KMT5A).
  • Supplied as a solid, soluble in DMSO for assay preparation.
  • Available in small research pack sizes suitable for preclinical studies.
Catalog No. 50-002-13849