Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Hydroxy-3-methoxy-5-nitrobenzoic acid | 15785-54-3 | 99.9% | C8H7NO6 | 50 G
SDP

Supplier:  Medchemexpress LLC HY7901250G

Encompass_Preferred

4-Hydroxy-3-methoxy-5-nitrobenzoic acid is a chemical compound used as a drug intermediate for the synthesis of various active compounds. This product is for research use only.

  • High purity of 99.9%
  • Solid appearance, light yellow to yellow color
  • Molecular weight of 213.15
  • Recommended storage at 4°C under nitrogen
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C under nitrogen

Catalog No. 50-004-34898


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.