Learn More
Medchemexpress LLC 4-Hydroxy-3-methoxyphenylglycol sulfate potassium | 71324-20-4 | 98.8% | 302.34 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11344110MG
Encompass_Preferred
4-Hydroxy-3-methoxyphenylglycol sulfate potassium is a biosynthesis product that releases Na2SO4 and free ethylene glycol on acidic hydrolysis.
Catalog No. 50-004-36593