Learn More
Medchemexpress LLC 4-hydroxy atorvastatin hemicalcium | 265989-44-4 | 96.2% | 594.68 g/mol | C33H35FN2O6 · 1/2 Ca | 5 MG

Supplier: Medchemexpress LLC HY1373105MG
4-Hydroxy atorvastatin hemicalcium is the hemicalcium salt of the para-hydroxy metabolite of atorvastatin. It is supplied as a research-grade reference standard for metabolite identification, pharmacokinetic, and analytical studies, offered in small pack sizes for laboratory use.
- Hemicalcium salt of the atorvastatin para-hydroxy metabolite.
- Provided as a research-grade reference standard.
- Supplied in a 5 mg pack suitable for analytical work.
- Purity 96.2% by HPLC.
- Molecular weight 594.68 g/mol and formula C33H35FN2O6 · 1/2 Ca.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.