Learn More
Medchemexpress LLC 4-hydroxy-N-[[4-(4-methyl-1,3-thiazol-5-yl)phenyl]methyl]pyrrolidine-2-carboxamide | 2097381-85-4 | MFCD30742947 | >=98.0% | 523.6 g/mol | C27H33N5O4S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10094725MG
Encompass_Preferred
VH-298 is a cell-active small-molecule inhibitor of the VHL:HIF-α interaction used as a research tool to stabilize HIF-α and investigate hypoxia signaling and targeted protein degradation. It is applicable to biochemical and cellular assays and is commonly used in PROTAC development.
  • Potent inhibitor of VHL:HIF-α interaction.
  • Cell-active; promotes HIF-α accumulation in cells.
  • Useful as a tool compound in hypoxia signaling studies and PROTAC research.
  • High purity suitable for biochemical and cellular assays.
  • Well-characterized chemical identifiers for compound tracking.
Catalog No. 50-002-12364