Learn More
Cayman Chemical 4-hydroxy Nonal Alkyn 100ug
SDP
Supplier:  Cayman Chemical 13265100 UG
Encompass_Preferred
A form of 4-HNE with a terminal alkyne; for use in linking reactions (click chemistry) for detection of 4-HNE-modified DNA and proteins
Catalog No. 50-295-2917