Learn More
Cayman Chemical 4-hydroxy Nonal-d3 500ug
SDP
Supplier:  Cayman Chemical 332101500 UG
Encompass_Preferred
An internal standard for the quantification of 4-HNE by GC- or LC-MS
Catalog No. 50-295-0357