Learn More
Cayman Chemical 4-hydroxy Valsartn 2.5mg
SDP
Supplier:  Cayman Chemical 278492.5 MG
Encompass_Preferred
A major metabolite of valsartan; reduces epinephrine and collagen-induced platelet aggregation in human whole blood
Catalog No. 50-293-8273