Learn More
Medchemexpress LLC 4-Hydroxyantipyrine | 1672-63-5 | 99.9% | 1 ML

Supplier: Medchemexpress LLC HYB215010MM/1ML
4-Hydroxyantipyrine is the primary metabolite of Antipyrine and functions as a biodistribution promoter. It enhances the distribution of Citicoline and Antipyrine in the brain by increasing their concentration ratio. In vitro studies show it increases the tissue-to-plasma concentration ratio of Citicoline in the brain and liver, and of thiopental sodium in the brain, liver, and heart. It also improves the blood-brain barrier (BBB) permeability of Antipyrine, linked to an increase in Antipyrine's Kp value in the brain. This product is for research use only.
- Purity: 99.9%
- Molecular weight: 204.23
- Chemical formula: C11H12N2O2
- Appearance: Solid
- Color: Light yellow to yellow
- Recommended storage for solution: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.