Learn More
Medchemexpress LLC 4-Hydroxybenzaldehyde | 123-08-0 | MFCD00006939 | 100.0% | 122.12 g/mol | C7H6O2 | 25mg
SDP
Supplier:  Medchemexpress LLC HYY0313R25MG
Encompass_Preferred
p-Hydroxybenzaldehyde (Standard) is the analytical standard of p-Hydroxybenzaldehyde This product is intended for research and analytical applications p-Hydroxybenzaldehyde is a one of the major components in vanilla aroma with antagonistic effect on GABAA receptor of the 1 2 2S subtype at high concentrations
Catalog No. 50-002-68209