Learn More
Medchemexpress LLC 4-hydroxybenzoic acid-13C | 146672-02-8 | MFCD01075595 | 99.2% | 139.11 | C6 13CH6O3 | 25mg
SDP
Supplier:  Medchemexpress LLC HYY0264S225MG
Encompass_Preferred
4-Hydroxybenzoic acid-13C is the 13C labeled 4-Hydroxybenzoic acid[1] 4-Hydroxybenzoic acid a phenolic derivative of benzoic acid could inhibit most gram-positive and some gram-negative bacteria with an IC50 of 160 g/mL[2]
Catalog No. 50-002-66507