Learn More
Medchemexpress LLC 4-hydroxybenzyl alcohol | 623-05-2 | MFCD00004658 | 99.8% | 124.14 | C7H8O2 | 100g
Supplier: Medchemexpress LLC HYY0892100G
4-Hydroxybenzyl alcohol is a phenolic compound widely distributed in various kinds of plants Anti-inflammatory anti-oxidant anti-nociceptive activity Neuroprotective effect Inhibitor of tumor angiogenesis and growth[1][2][3][4]
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.