Learn More
Medchemexpress LLC 4-hydroxybenzyl alcohol | 623-05-2 | MFCD00004658 | 98.5% | 124.14 | C7H8O2 | 25mg
SDP
Supplier:  Medchemexpress LLC HYY0892R25MG
Encompass_Preferred
4-Hydroxybenzyl alcohol (Standard) is the analytical standard of 4-Hydroxybenzyl alcohol This product is intended for research and analytical applications 4-Hydroxybenzyl alcohol is a phenolic compound widely distributed in various kinds of plants Anti-inflammatory anti-oxidant anti-nociceptive activity Neuroprotective effect Inhibitor of tumor angiogenesis and growth[1][2][3][4]
Catalog No. 50-002-68522
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.