Learn More
Medchemexpress LLC 4-hydroxybenzyl cyanide | 14191-95-8 | MFCD00002383 | 98.0% | 133.15 g/mol | C8H7NO | 1 ML

Supplier: Medchemexpress LLC HYW01582310MM/1ML
4-Hydroxybenzyl cyanide is an endogenous metabolite provided as an analytical reference standard for qualitative and quantitative research. It is commonly used in method development and calibration for HPLC, GC, and MS.
- Analytical reference standard for HPLC, GC, and MS applications.
- High purity: 98.0%.
- Molecular formula C8H7NO; molecular weight 133.15 g/mol.
- Available as a solid in multiple sizes (10 mg-5 g).
- Recommended storage: powder at -20°C (long term); in solution at -80°C (short term).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.