Learn More
Selleck Chemical LLC 4-Hydroxychalcone 25mg 20426-12-4 P-Cinnamoylphenol
SDP
Supplier:  Selleck Chemical LLC S476325MG
Encompass_Preferred
4-Hydroxychalcone (P-Cinnamoylphenol) is a chalcone metabolite with diverse biological activities. It inhibits TNF?-induced NF-?B pathway activation in a dose-dependent manner and also activates BMP signaling. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8332