Learn More
Medchemexpress LLC 4-hydroxydiphenylamine | 122-37-2 | MFCD00020142 | 98.0% | 185.22 g/mol | C12H11NO | 1 ML

Supplier: Medchemexpress LLC HYW08832010MM/1ML
4-Hydroxydiphenylamine is supplied as a 10 mM solution in DMSO (1 mL) for use in biochemical and pharmacological research. The compound has CAS number 122-37-2, molecular formula C12H11NO, and a molecular weight of 185.22 g/mol. Typical purity is 98.0%.
- 10 mM solution in DMSO, 1 mL vial.
- CAS number 122-37-2.
- Molecular formula C12H11NO.
- Molecular weight 185.22 g/mol.
- Purity approximately 98.0%.
- Intended for research use in biochemical assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.