Learn More
Selleck Chemical LLC 4-Hydroxyestrone E8109-1MG
SDP
Supplier:  Selleck Chemical LLC E81091MG
Encompass_Preferred
4-Hydroxyestrone (4-OHE1) an estrone metabolite has strong neuroprotective effect against oxidative neurotoxicity 4-Hydroxyestrone increases cytoplasmic translocation of p53 resulting from SIRT1-mediated deacetylation of p53 4-Hydroxyestrone has little estrogenic activity
Catalog No. 50-000-41875