Learn More
Medchemexpress LLC 4'-Hydroxyflavanone | 6515-37-3 | MFCD00006734 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN84955MG
Encompass_Preferred
4'-Hydroxyflavanone is an inhibitor of SREBP maturation and lipid synthesis. It is a synthetic analogue of flavanone and has potential for hepatic steatosis and dyslipidemia research.
  • Inhibitor of SREBP maturation
  • Inhibitor of lipid synthesis
  • Synthetic analogue of flavanone
  • Potential for hepatic steatosis research
  • Potential for dyslipidemia research
Catalog No. 50-004-17502