Learn More
Medchemexpress LLC 4-Hydroxyindole (Standard) | 2380-94-1 | 97% | 133.15 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY34596R50MG
Encompass_Preferred
4-Hydroxyindole (Standard) is the analytical standard of 4-Hydroxyindole, intended for research and analytical applications. It is a hydroxyindole that is 1H-indole substituted by a hydroxy group at position 4. This compound is an important raw material or intermediate in the synthesis of pharmaceutical products and industrial polymers.
  • Used as an analytical standard
  • Reference standard supplied assay
  • Commonly used in qualitative research experiments
  • Commonly used in quantitative research experiments
  • Commonly used in methodological research experiments
  • Applicable in HPLC, GC, and MS
Catalog No. 50-004-04331